Synonyms:
4-Methylbenzenesulphonamide
, 4-Methylbenzenesulfonamide
, p-Toluenesulfonamide
, p-Toluenesulphonamide
T0281-25G
T0281-500G
TCT0281-500GEA
37.53
USD
InStock
TCT0281-500G
TCT0281-025G
Toluene-4-sulfonamide ≥98.0% (by HPLC, total nitrogen)
Toluene-4-sulphonamide
CAS Number: 70-55-3
MDL Number: MFCD00011692
Molecular Formula: C7H9NO2S
Molecular Weight: 171.21
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Boiling point (°C): 214
Melting point (°C): 137
Flash Point (°C): 202
Other Product Names / Synonyms: PASAM