Synonyms:
Tribromomethane
Stabilization:
Stabilised with Stabilised
T0348-500G
T0348-25G
TCT0348-500GEA
143.66
USD
InStock
TCT0348-500G
TCT0348-25G
Bromoform ≥98.0% stabilized
Bromoform
(stabilized with Ethanol)
CAS Number: 75-25-2
MDL Number: MFCD00000128
Molecular Formula: CHBr3
Molecular Weight: 252.73
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 149
Melting point (°C): 8
Specific Gravity (20/20): 2.62
Other Product Names / Synonyms: Tribromomethane