Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriethylsilane ≥97.0%

Supplier: TCI America
Synonyms: Triethylchlorosilane

T0589-5G T0589-25G T0589-100G
TCT0589-025GEA 74.42 USD InStock
TCT0589-025G TCT0589-005G TCT0589-100G
Chlorotriethylsilane ≥97.0%
Chlorotriethylsilane
CAS Number: 994-30-9
MDL Number: MFCD00000507
Molecular Formula: C6H15ClSi
Molecular Weight: 150.72
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 149
Flash Point (°C): 30
Specific Gravity (20/20): 0.90

Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Triethylsilyl Chloride, Triethylchlorosilane, TES-Cl


Formula: C₆H₁₅ClSi
Boiling Pt: 149 °C
Density: 0.90 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/91/28994791.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty