Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriphenylsilane ≥95.0%

Supplier: TCI America
Danger
Synonyms: Triphenylchlorosilane

T0939-5G T0939-25G
TCT0939-025GEA 93.24 USD InStock
TCT0939-025G TCT0939-5G
Chlorotriphenylsilane ≥95.0%
Chlorotriphenylsilane
CAS Number: 76-86-8
MDL Number: MFCD00000496
Molecular Formula: C18H15ClSi
Molecular Weight: 294.85
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 208
Melting point (°C): 96
Flash Point (°C): 200

Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Chlorotriphenylsilane, Triphenylsilyl Chloride


Formula: C₁₈H₁₅ClSi
MW: 294.86 g/mol
Boiling Pt: 208 °C
Melting Pt: 96 °C
Density: 1.0165 g/cm³ (20 °C)
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00000496
CAS Number: 76-86-8
EINECS: 200-989-0
UN: 3261
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/50/30/28995030.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty