Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluamide ≥98.0%

Supplier: TCI America
Synonyms: 4-Methylbenzamide

T1257-25G
TCT1257-025GEA 32.48 USD InStock
TCT1257-025G
p-Toluamide ≥98.0%
p-Toluamide
CAS Number: 619-55-6
MDL Number: MFCD00008002
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 159

Other Product Names / Synonyms: 4-Methylbenzamide


Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 159 °C
MDL Number: MFCD00008002
CAS Number: 619-55-6
EINECS: 210-600-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/52/98/28995298.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty