Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylammonium hexafluorophosphate

Supplier: TCI America
Synonyms: Tetramethylammoniumhexafluorophosphate

T1296-25G
TCT1296-025GEA 67.99 USD InStock
TCT1296-025G
Tetramethylammonium hexafluorophosphate
Tetramethylammonium hexafluorophosphate
CAS Number: 558-32-7
MDL Number: MFCD00012012
Molecular Formula: C4H12F6NP
Molecular Weight: 219.11
Form: Crystal
Color: White


Formula: C₄H₁₂F₆NP
MW: 219.11 g/mol
Melting Pt: 300 °C
Density: 1.617 g/cm³ (25 °C)
MDL Number: MFCD00012012
CAS Number: 558-32-7
EINECS: 209-194-3
UN: 2923
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/33/28995333.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty