Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,2-Tribromoethanol ≥98.0%

Supplier: TCI America
Warning
Synonyms: Tribromoethyl alcohol

T1420-25G
TCT1420-025GEA 118.72 USD InStock
TCT1420-025G
2,2,2-Tribromoethanol ≥98.0%
2,2,2-Tribromoethanol
CAS Number: 75-80-9
MDL Number: MFCD00004671
Molecular Formula: C2H3Br3O
Molecular Weight: 282.76
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 94
Melting point (°C): 81
Flash Point (°C): 110


Formula: Br₃CCH₂OH
MW: 282.76 g/mol
Boiling Pt: 94 °C
Melting Pt: 81 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9
EINECS: 200-903-1
Merck Index: 12,09743
https://us-legacy.vwr.com/stibo/web/std.lang.all/54/40/28995440.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty