Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Thiophenemethanol ≥96.0%

Supplier: TCI America
Synonyms: Thiophen-3-methanol 3-Thienylmethanol

T1660-25G T1660-5G
TCT1660-25GEA 322.77 USD InStock
TCT1660-25G TCT1660-005G
3-Thiophenemethanol ≥96.0%
3-Thiophenemethanol
CAS Number: 71637-34-8
MDL Number: MFCD00014534
Molecular Formula: C5H6OS
Molecular Weight: 114.16
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 73
Flash Point (°C): 101
Specific Gravity (20/20): 1.22

Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Hydroxymethylthiophene


Formula: C₅H₆OS
MW: 114.17 g/mol
Boiling Pt: 73 °C
Density: 1.22 (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00014534
CAS Number: 71637-34-8
EINECS: 275-741-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/56/50/28995650.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty