Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triacetoxymethylsilane ≥90.0%

Supplier: TCI America
Synonyms: Triacetoxy(methyl)silaneMethylsilanetriyl triacetate

T1786-25G
TCT1786-025GEA 39.55 USD InStock
TCT1786-025G
Triacetoxymethylsilane ≥90.0%
Triacetoxymethylsilane
CAS Number: 4253-34-3
MDL Number: MFCD00008694
Molecular Formula: C7H12O6Si
Molecular Weight: 220.25
Purity/Analysis Method: 90.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 85
Freezing point (°C): 40

Other Product Names / Synonyms: Methyltriacetoxysilane


Formula: C₇H₁₂O₆Si
MW: 220.25 g/mol
Flash Pt: 85 °C
MDL Number: MFCD00008694
CAS Number: 4253-34-3
EINECS: 224-221-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/57/63/28995763.jpg
Order Now
Specification Test Results
Purity/Analysis Method 90.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty