Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl valerate ≥99.0%

Supplier: TCI America
Synonyms: Valeric acid methyl ester Methyl Pentanoate

V0005-25ML V0005-500ML
TCV0005-025MLEA 23.35 USD InStock
TCV0005-025ML TCV0005-500ML
Methyl valerate ≥99.0%
Methyl valerate
CAS Number: 624-24-8
MDL Number: MFCD00009478
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 128
Flash Point (°C): 29
Specific Gravity (20/20): 0.89

Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Valeric Acid Methyl Ester


Formula: C₆H₁₂O₂
Boiling Pt: 128 °C
Melting Pt: –91 °C
Density: 0.89 (20 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00009478
CAS Number: 624-24-8
EINECS: 210-838-0
UN: 3272
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/59/28997259.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty