|
General Protection
Clothing that protects you in wide array of applications and environments from routine industrial to biopharmaceutical and aerospace. The ProShield® brand has broad offering to suit almost every requirement. |
|
DuPont™ Tyvek® 400
- Composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont
- Provides an ideal balance of protection, durability and comfort of any limited use fabric technology
- Offers an inherent barrier against particles (down to 1.0 micron in size)
- Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Learn more
|
|
DuPont™ Tyvek® 400 D
- Combines the protection, durability and comfort of DuPont™ Tyvek® fabric on the front and the comfort, softness and breathability of DuPont™ ProShield® 10 fabric on the back
- Suitable for applications where comfort requirements are combined with limited protective requirements for frontal exposures
- Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance
Learn more
|
|
DuPont™ Tyvek® 500
- Composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont
- Provides an ideal balance of protection, durability and comfort.
- Available with external serged seams that provide the performance of Type 5/6 garments, which have been tested to standards against light liquid aerosols and airborne solid particles
- Provides an effective barrier against particles (down to 1.0 micron in size)
- Suitable for applications such as pharmaceutical handling, chemical processing, automatic spray painting, maintenance and many others.
Learn more
|
|
DuPont™ Tyvek® 600
- Composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont
- Provides an ideal balance of protection, durability and comfort
- Available with serged and over-taped seams that provides the performance of Type 4/5/6 garments, which have been tested to standards against heavy liquid aerosols and airborne solid particles
- Provides an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size)
- Suitable for applications such as pharmaceutical manufacturing, medical applications, research and biosecurity laboratories, nuclear and maintenance
Learn more
|
|
DuPont™ Tyvek® 800
- Combines resistance to low-concentration, water-based, inorganic chemicals (even under pressure) with the strength and durability of Tyvek® thanks to an innovative fabric technology and enhanced garment design
- Available with serged and over-taped seams that provide the performance of Type 3/4/5/6 garments, which have been tested to standards against pressurized liquids, heavy liquid aerosols and airborne solid particles
- Provides an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size), as well as oil repellency
- Suitable for applications such as industrial cleaning, chemical packaging and redistribution, waste treatment and disposal, and environmental remediation
Learn more
|
|
DuPont™ ProShield® 10
- Spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont
- Optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments
- Designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications
- Available in a variety of coverall styles in a choice of white, blue and gray
- Applications include: general maintenance, janitorial/cleaning and other dirty work
Learn more
|
|
DuPont™ ProShield® 50
- Suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications
- Made with a microporous film that is laminated to a spundbonded polypropylene substrate
- Applications include: janitorial, sanitation, and general industrial maintenance
Learn more
|
|
DuPont™ ProShield® 60
- Suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications
- Made with a microporous film that is laminated to a spundbonded polypropylene substrate
- Applications include: automotive refinishing, waste cleanup and remediation
Learn more
|
|
DuPont™ ProShield® 70 Shoe and Boot Covers
- ProShield® 70 fabric, used exclusively for shoe and boot cover products, is a spunbonded polypropylene substrate that is coated with a low-density polyethylene film for enhanced skid resistance and barrier performance