>
More...
>
Events
>
Avantor Webinars
>
Improve Your Biochromatography Separations - How to Optimize your Charge Variant Analysis
Improve Your Biochromatography Separations - How to Optimize your Charge Variant AnalysisWEBINARReplayTaking charge of variant analysisThe analysis of charged variants is a regulatory requirement for bio-therapeutic proteins. Ion exchange chromatography is useful in the analysis of enzymatic post-translational modifications and chemical modifications that can occur on a bio-therapeutic. Understanding column selection (WCX vs SCX) and buffer optimization (salt vs pH gradients) is key to developing a robust, high resolution, and sensitive assay. Join us to hear about optimising ion exchange method development, best practices, and the latest innovations of coupling charged variant analysis to mass spectrometry (CVA-MS). Presented by: Dr. Steve Guilfoyle Steve Guilfoyle, a former research physicist, joined the Thermo Fisher team as a biopharmaceutical and pharmaceutical market development manager. Currently, as a training specialist his passion and experience is empowering individuals with the tools to optimize productivity and performance: enabling researchers to solve their bio-chromatographic challenges. |