Would you like to log in again?
AtmoSense™ Oxygen Monitor, 115 V
Supplier: Labconco
5255500
10002-752EA 4520 USD InStock
10002-752
AtmoSense™ Oxygen Monitor, 115 V
Fume Hoods and Enclosures Chemical Fume Hoods
The AtmoSense™ Oxygen monitor is designed for general purpose trace oxygen detection as low as 1 ppm.
Designed for placement atop transfer chamber of a Labconco® Protector® controlled atmosphere, combination glove box, or the Precise® controlled atmosphere glove box rather than inside. Operates in-line with the AtmosPure™ Re-Gen gas purifier. Constructed with a durable glacier white powder-coated housing. Gas inlet and outlet connections are 1.9 cm (³/₄") brass compression fittings.
Equipped with three user-configurable oxygen alarm levels and three relays. Also provided with 4 to 20 mA output signal with 0 to 5 V output signal for transmission of live data to a user-supplied personal computer.
The device provides a digital LCD readout from 0 to 1000 ppm oxygen, with resolution accuracy of 0.1 ppm from 0 to 200 ppm and a resolution accuracy of 1 ppm from 200 to 1000 ppm. Includes certificate of initial calibration between 90 to 199 ppm.
Ordering information: The AtmoSense™ Oxygen monitor must be used with circulation tubing kit 5441808 (sold separately). The AtmoSense™ Oxygen monitor includes 100 to 115 V cord with IEC 320 C14 plug. With 12 V DC input voltage for operation on 50/60 Hz powered from 100, 115, or 208/240 V AC. Power cords with other plug types are available (sold separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty