Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
CApture™ BT Fuming Chambers, Labconco
Supplier: Labconco
3170003 3182900 3170004 3170001 3170005 3170002 3170000
10014-346EA 2210 USD InStock
10014-346 10014-336 10014-338 10014-200 10014-196 10014-340 10014-198
CApture™ BT Fuming Chambers, Labconco
Fume Hoods and Enclosures Forensic Enclosures
The CApture™ BT is a spacious benchtop Cyanoacrylate (CA) evidence fuming chamber with a durable, easy-to-clean stainless steel liner
The microprocessor-controlled operating system allows for adjustment of chamber humidity to 80% +/- 5% relative humidity, pre-fuming humidity incubation, fuming temperature up to 424°F within +/- 1%, and fuming and exhaust times. System stores 20 programs, has 3 factory set programs (Low, High, Lumincyano) and includes diagnostic mode with audible/visual alerts for program cycle completion. Acti-Vent™ Smart Controls purge air during critical steps to protect evidence from over fuming exposure and the user from fumes.
The CApture™ BT is constructed of a powder-coated steel exterior, type 304 brushed stainless steel interior with removable perforated floor, four stainless steel hanging rods (26.9" long x 3/8" dia.) and quick-release sidewall brackets mounted at four heights. A storage drawer (28Wx13Dx1.5H") with flush-mounted handle is located beneath the chamber.
It features a built-in humidifier, full-color eye-level LCD control screen, recirculation blower, exhaust blower and interior LED light. The tempered safety glass door has an electronic door lock to prevent opening during the fuming cycle and includes a power outage override. Filters are accessible from inside the chamber for easy replacement.
Base stand adds portability and storage to CApture™ BT Fuming Chamber. Powder-coated steel frame with 5" diameter, toe-locking, polyurethane casters. Features shelves and compartment to hold supplies and accessories. Stand height is manually adjustable from 21" to 28". Weight capacity of the stand is 400 lbs."
Its ample interior (19 cubic feet with 58.6" diagonal length) with shelving system maximizes fuming capacity and accommodates large evidence including up to 15 long guns.
Certifications: ETL and ETL-C listed. CE conformity marking.
Ordering information: Four leveling feet, 10' power cord and plug, one prefilter, one carbon filter and one humidifier filter are included with each CApture™ BT Fuming Chamber. Optional accessories include mobile base stand, shelves, casing and long gun holders, hanging rods, clips and security tags (each sold separately).
Super Glue, fuming dishes, and purified water are required (not included).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty