Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
PuriCare® Open Access Station, Labconco®
Supplier: Labconco
PuriCare®
3820011 3820001 3820038 3820020 3820024 3820033 3820026 3820023 3820034 3820021 3820036 3820028 3820000
10033-454EA 19510 USD InStock
10033-454 10033-322 10033-310 10033-320 82003-762 82003-760 10033-318 10033-328 10033-316 10033-326 10033-314 10033-324 10033-312
PuriCare® Open Access Station, Labconco®
Fume Hoods and Enclosures Animal Research Enclosures
PuriCare Open Access Stations offer the utmost efficiency for cage changing operations
The PuriCare Open Access Station is supplied with two 99.99% efficient HEPA filters, disposable inlet and exhaust prefilters, stainless steel dished work surface, polyurethane casters, as well as ADA-compliant light and blower switches, electrical duplex, and two Minihelic® II pressure gauges. Unit features an attractive, durable glacier white and slate gray epoxy-coated steel exterior. It also features a built-in electric hydraulic lift that allows the working height to be adjusted from 93–114cm (36.5–45.0") with the push of a button.
Open access on two sides provides ample workspace for one or two operators to transfer animals from dirty cages to clean ones. The station provides personnel, animal, and environmental protection as air passes through prefilters and HEPA filters. HEPA filters provide ISO Class 5 (FED-STD-209E Class 100/M3.5) clean air in the work area.
Certifications: Conform to: UL Standard 3101/61010-1 (115 volt model), CAN/CSA C22.2 No. 1010.1-92 (115 volt model), CE Conformity Marking (230 volt model).
Ordering information: Supplied with a three-year manufacturer’s limited warranty on parts and labor.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty