Would you like to log in again?
Protector® PVC Perchloric Acid and Acid Digestion Laboratory Hoods, Labconco®
Supplier: Labconco
141410001 140410001 140510020 140510000 141410021 140610002 140410000 141610021 141410002 140810000 140410021 140510002 141810020 141610020 140610000 140810002 141810002 141610000 141510000 141510020 141610001 140410002 140810020 140510021 140510001 141610002 141410020 141810021 140610021 141810001 140810001 141810000 140610001 141510002 141510021 140810021 140410020 141510001 140610020 141410000
10118-518EA 28605 USD InStock
10118-518 10118-512 10118-534 10118-556 10118-554 10118-510 10118-532 10118-538 10118-516 10118-514 10118-536 10118-482 10118-480 10118-486 10118-520 10118-542 10118-484 10118-540 10118-508 10118-468 10118-502 10118-546 10118-524 10118-488 10118-522 10118-544 10118-500 10118-528 10118-506 10118-526 10118-548 10118-504 10118-494 10118-492 10118-552 10118-530 10118-498 10118-550 10118-496 10118-490
Protector® PVC Perchloric Acid and Acid Digestion Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Recommended whenever perchloric acid is used in combination with other corrosive inorganic reagents.
Interior and integral bottom with drainage trough are constructed of heat-welded Type I unplasticized polyvinyl chloride (PVC).
Perchloric acid hoods feature tempered safety glass sash. Acid digestion hoods utilize LEXAN® plastic sash to resist hydrofluoric acid exposure. Hoods have a glacier white powder-coated steel exterior.
Hoods are factory prepared for up to eight service fixtures and four electrical duplexes. They are supplied with two pre-plumbed service fixtures with forged brass valves; lower right side with brass tubing and plastic serrated hose connector for gas, and lower left side with gooseneck faucet for cold water. Components for converting gas fixture to air and vacuum are provided. Hoods have a 32.4 cm (123/4") O.D. exhaust collar.
A black, enamel-coated hardboard surface supports the integral work surface. Built-in washdown system with internal piping and spray nozzles facilitates removal of hazardous chemical residue from hood interior and ductwork. Pre-wired LED lighting rated to 50000 hours gives a bright, nonglare working surface. The 73.6 cm (29.1") interior depth permits use of oversize apparatus.
Certifications: ETL and ETL-C listed.
Accessories information: Fabricated from vacuum formed unplasticized Type 1 polyvinyl chloride (PVC) and made specifically to handle extremely corrosive atmospheres, these blowers provide optimum chemical resistance to high order oxidizers such as perchloric acid.PVC blowers have a dry powder epoxy-coated base and weather cover for complete protection in extreme weather. Impellers are made of injection molded PVDF to resist highly corrosive atmospheres.
Ordering information: The 4' to 6' models are available with 2 service fixtures and 1 electrical receptacle included. The 8' models are available with 2 service fixtures and 2 electrical receptacles included. A dedicated, remotely located exhaust blower and a base cabinet or stand are required for operation (all sold separately).
Order Now
ORDER
ACCESSORIES
SERVICES
Services: VWRCATALYST™ offers customer on-site validation, testing and certification services including face velocity profile test, smoke generation test, tracer gas test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST@vwr.com for more details. You can also visit https://vwr.com/equip_services.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty