Would you like to log in again?
WATERPRO WALL MOUNTING PANEL KIT
Supplier: Labconco
WaterPro Wall Mounting Panel Kit^ Includes stainless steel panel and hardware for mounting a WaterPro RO or WaterPro PS System to a flat wall surface, Dimensions: 38in w x 10in d x 37in h, Stands support WaterPro Purification Systems on horizontal surfaces when wall mounting is not desired.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WATERPRO WALL MOUNTING PANEL KIT 10118-964 9078800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty