Would you like to log in again?
EXPAND CHMBR KIT CAPTUR PRTBL FUMING SYS
Supplier: Labconco
Expandable Fuming Chamber Kit for Capture^ Includes a carrying case that holds many portable, configurable frame pieces, clear plastic bags (used to form the chamber), and a portable fan to evenly distribute fumes, Dimensions: 24in l x 12in w x 12in h
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EXPAND CHMBR KIT CAPTUR PRTBL FUMING SYS 10118-966 3427000 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty