Would you like to log in again?
Intelli-Sense™ Multi-Speed Coated Steel Blowers, Labconco
Supplier: Labconco
Intelli-Sense™
7061122 7061110 7061112 7061120 7061123 7061113 7061121 7061111
10127-920EA 4090 USD InStock
10127-920 10127-922 10127-924 76330-110 10127-926 76330-104 76330-106 76330-108
Intelli-Sense™ Multi-Speed Coated Steel Blowers, Labconco
Ventilators
Intelli-Sense™ Multi-Speed Coated Steel Blowers are recommended for low to moderately corrosive applications
They feature a direct drive ECM motor, which consumes one third less energy and is easier and less expensive to maintain than belt drive motors, and a phenolic-coated impeller and housing. Unit comes with control box and 3 speed blower switch. Adjustable over broad range of airflows.
The Intelli-Sense Blower provides an economical alternative to traditional on/off blowers. Users may close the sash and turn the Intelli-Sense Blower switch to standby mode so that the hood operates at a lower velocity yet fulfills minimum air exchange requirements. Lower velocity modes have the added benefit of quieter operation than full velocity mode.
For greater corrosion protection, refer to the Fiberglass Blowers and PVC Blowers. Phenolic coating on impeller and housing is corrosion resistant. Powder-coated base and weather cover provide long life in extreme weather.
Ordering information: Models without Control Box are only for use with Labconco Protector I-S series Fume Hoods.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty