Would you like to log in again?
KNA 6 UNIT DIGESTION RACK 230V 60HZ 3PH
Supplier: Labconco
Digestion Apparatus^ Six-Place Kjeldahl Digestion Apparatus Dimensions: 44in w x 23in d x 62in h Electrical: 230 V, three phase, 60 Hz, 11 amps
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KNA 6 UNIT DIGESTION RACK 230V 60HZ 3PH 10129-228 2125613 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty