Would you like to log in again?
WORKSURFACE 2' X 29"D SST
Supplier: Labconco
Work Surface^ Work Surface, Type 304 Stainless Steel, 24x29x0.8 inch, 710 lbs, Applicable Products: XPert Filtered Balance Stations, Xpert Filtered Balance Systems, Hood, Enclosure or Glove Box Width: 2 inch.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WORKSURFACE 2' X 29"D SST 10147-152 3970602 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty