Would you like to log in again?
MICROCENTRIFUGE TB HLDR 12WELL
Supplier: Labconco
Holder^ Microcentrifuge Tube Holder, Anodized aluminum block with 12 bore holes, accommodates 1.7 ml microcentrifuge tubes, placed on any flat freeze drying surface, built-in vacuum drying chamber of FreeZone Console Systems, or on Product, Heated Product, FreeZone or Bulk Tray Dryer Shelves
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICROCENTRIFUGE TB HLDR 12WELL 10148-016 7439300 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty