Would you like to log in again?
CLEAR STOPPERING CHMBR 230V
Supplier: Labconco
Clear Stoppering Chamber^ Clear Stoppering Chamber, 15V, 50/60 Hz. For use with 6L, 12L and 18L Freeze Dryers (freeze dry system or contain samples in bulk), Weight: 30.0 lbs, Weight metric: 13.6 kg, Dimensions: 23.0inch x 13.5inch diameter, Electrical: 230 volts, 50/60 Hz, Domestic
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEAR STOPPERING CHMBR 230V 10148-022 7868030 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty