Would you like to log in again?
LIQUID NITROGEN TRAP
Supplier: Labconco
Acid Trap Insert^ Acid Trap Insert, Glacier white powder-coated steel construction, 2 valves may be opened (trap) or closed (by-pass). 3/4inch+ OD inlet and outlet connections, 20inch wire-reinforced PVC tubing, 3/4inch Inner diameter; and 2 clamps. Liquid nitrogen required
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LIQUID NITROGEN TRAP 10148-026 7394800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty