Would you like to log in again?
FLASK SEAL LYPH-LOCK 25 & 50ML
Supplier: Labconco
Flask Seal^ Lyph-Lock Flask Seal, Flexible silicone rubber seal grasps the top and bottom of the Lyph-Lock flask, ring holds securely together during pre-freezing and lyophilization, Weight: 0.5 lbs, Weight metric: 0.2 kg, Volume: 25-50 ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLASK SEAL LYPH-LOCK 25 & 50ML 10148-036 7559000 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty