Would you like to log in again?
STAINLESS STEEL FLASK ADAPTER 29/32
Supplier: Labconco
Adapter^ Stainless Steel Adapter, allows connection of 29/32 round bottom flasks to valve ports on drying chambers and manifolds
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STAINLESS STEEL FLASK ADAPTER 29/32 10148-044 7543800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty