Would you like to log in again?
GLASS LID SMALL FOR 4.5L FD
Supplier: Labconco
Glass lid^ Glass lid, replaces the acrylic lid included with FreeZone 4.5 Liter Freeze Dry System with built-in 10-port drying chamber, Provides additional protection from solvents, that attack acrylic, such as acetonitrile, Weight: 2.5 lbs, Weight metric: 1.1 kg, Dimensions: 10.12inch diameter
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLASS LID SMALL FOR 4.5L FD 10148-054 7439903 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty