Would you like to log in again?
TISSUE SCREEN KIT, 5' PREMIER
Supplier: Labconco
Tissue Screen Kit 5Feet^ Tissue Screen Kit, Mount behind the baffles of a chemical fume hood for trapping tissue wipers, other small, loose materials which may be caught in the airflow, Includes: Mounting hardware, powder-coated SS screen, For Protector P
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TISSUE SCREEN KIT, 5' PREMIER 10216-838 9577111 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty