Would you like to log in again?
Protector® XL™ Floor-Mounted Laboratory Hoods with Horizontal Sashes, Labconco
Supplier: Labconco
112126002 112136002 112816002 112236002 112036002 112216002 112026002 112836002 112226002 112016002 112826002 112116002
10222-160EA 35275 USD InStock
10222-160 10221-904 10222-162 10221-902 10222-164 10222-166 10222-154 10221-906 10222-168 10222-156 10222-158 10221-900
Protector® XL™ Floor-Mounted Laboratory Hoods with Horizontal Sashes, Labconco
Fume Hoods and Enclosures Chemical Fume Hoods
Protector XL floor-mounted laboratory hoods with horizontal-sliding sashes are suitable for general chemistry applications and have the dimensions required for over-sized equipment.
These hoods are constructed of durable, powder-coated steel with four tempered safety glass sashes and a corrosion- and chemical-resistant, fiberglass-reinforced composite panel liner that surpasses all national codes for flame spread. Features include removable access panels, 73.5" loading height and pre-wired LED lighting rated for 50000 hours.
Laboratory workers with applications requiring distillation racks, extraction apparatus or large drums and carboys benefit from the increased loading height these hoods provide.
Certifications: Protector XL floor-mounted laboratory hoods with horizontal-sliding sashes are performance tested to ASHRAE 110-1995, ETL-listed and CE approved. See our brochure for a complete list of regulations and standards to which these models conform.
Ordering information: All models require a remote blower and ductwork, not included. Models are available in widths of 8', 10', 12' and 16' and depths of 37.7", 43.7" and 55.7". These hoods can be ordered with 0 or 2 service fixtures, or 2 service fixtures and 2 GFCI Duplex receptacles. Electrical options of 100 to 115 volts, 50/60 Hz, 10 A or 208 to 230 volts, 50/60 Hz, 5 amps are offered. Hoods are shipped in multiple sections: top, bottom, and sashes. Protector XL floor-mounted hoods with vertical-rising sashes are also available. Contact your VWR representative for more information.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty