Testing confirms the Protector XL floor-mounted laboratory hoods with vertical-rising sashes meet the SEFA-1 standard of a low velocity, high performance hood and may be operated as low as 60 fpm.
These hoods are constructed of durable, powder-coated steel with two tempered safety glass sashes and a corrosion- and chemical-resistant, fiberglass-reinforced composite panel liner that surpasses all national codes for flame spread. Features include removable access panels, 63.6" loading height and pre-wired LED lighting rated for 50000 hours.
Protector XL floor-mounted laboratory hoods with vertical-rising sashes are suitable for general chemistry applications and accommodate laboratory procedures that require over-sized equipment or maximum working height.
Certifications: Protector XL floor-mounted laboratory hoods with vertical-rising sashes are performance tested to ASHRAE 110-1995, ETL-listed and CE approved. See our brochure for a complete list of regulations and standards to which these models conform.
Ordering information: All models require a remote blower and ductwork, not included. Models are available in widths of 4', 5', 6' and 8' and depths of 37.7", 43.7" and 55.7". These hoods can be ordered with 0 or 2 service fixtures, or 2 service fixtures and GFCI Duplex. 8' models with GFCI Duplex have 2 duplex receptacles, 1 on each side. Electrical options of 100 to 115 volts, 50/60 Hz, 10 A or 208 to 230 volts, 50/60 Hz, 5 amps are offered. Hoods are shipped in multiple sections: top, bottom, and sashes. Protector XL floor-mounted laboratory hoods with horizontal-sliding sashes are also available. Contact your VWR representative for more information.