Would you like to log in again?
REAR FINISH PANEL-8' XL FLRMT
Supplier: Labconco
Panel^ Glacier white powder-coated steel panel hides hood's interior components on the rear to provide finished appearance, Rear finish, 8inch, XL, Floormat, Recommended when hood is mounted on a casework island with back exposed. Adds 1.5inch to overall depth of Floor-Mount hoods.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REAR FINISH PANEL-8' XL FLRMT 10662-154 9548800 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty