Would you like to log in again?
Protector® Echo™ Filtered Fume Hood, Labconco
Supplier: Labconco
180510001 181410000 182810002 180410021 181410002 180610001 181410001 181610002 182610002 180410001 180810002 182410020 180610000 182410002 182510000 181510000 180610021 180510002 182810021 181610021 180510000 182810020 181510002 9577400 181410020 181610020 182410000 182610020 181610001 181810021 181510020 182510021 181810001 180810021 182810001 181810000 180610020 180410000 181810020 180410020 180510021 182410021 182810000 182610001 181810002 182610021 181510021 180610002 180810000 180810001 181610000 182510020 182510002 180510020 180810020 182510001 181510001 182610000 180410002 181410021 182410001
10746-860EA 32505.07 USD InStock
10746-860 10746-896 10746-852 10746-930 10746-776 10746-894 10746-850 10746-812 10746-810 10746-854 10746-932 10746-778 10746-816 10746-814 10746-858 10746-818 10746-870 10746-864 10746-900 10746-824 10746-822 10746-866 10746-904 10746-828 10746-902 10746-908 10746-906 10746-882 10746-880 10746-830 10746-874 10746-872 10746-834 10746-878 10746-912 10746-876 10746-910 10746-838 10746-916 10746-836 10746-914 10746-918 10747-112 10747-114 10747-116 10746-890 10746-842 10746-840 10746-884 10746-846 10746-802 10746-920 10746-800 10746-844 10746-888 10746-926 10746-806 10746-924 10746-804 10746-848 10746-808
Protector® Echo™ Filtered Fume Hood, Labconco
Fume Hoods and Enclosures Ductless Fume Hoods
The Protector® Echo™ Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Filter with the quality, innovation and energy-efficient fume hood designs of Labconco's unparalleled engineering to create an adaptable fully-featured fume hood that requires NO DUCTING! The Protector® Echo™ Filtered Fume Hood features many defining characteristics of Labconco Protector Hoods including the patented Eco-Foil™ Air Foil with Clean-Sweep openings, fiberglass-reinforced panel liner, fully closing sash, electrical receptacles and service fixtures.
GFH Filtration Technology allows the Protector Echo Filtered Fume Hood to perform well beyond the SEFA 9-2010 DH III definition. User safety is maintained through numerous built-in features that constantly monitor and prevent unsafe conditions without relying on user practices or modifications. These safeguards include controlled access to specified users via radio frequency identification (RFID) cards, a sensor package that detects primary filter breakthrough of solvent and acid fumes, laboratory air quality, sash position and temperature, audio/visual alarms, and intelligent filter indication of type and status to prevent saturated filters being re-installed. The optional gGuard® communication software offers real time monitoring of data.
Because the Protector Echo requires no connection to ductwork, initial building infrastructure change costs are minimized and allows the hood to be placed in hard-to-duct areas such as the center or bottom level of multi-level buildings. Should lab configurations change, the Protector Echo may be relocated without disrupting existing heating and air conditioning systems. The Neutrodine Filter has been tested and found efficient on over 500 chemicals, and may last 2 years or more before requiring replacement. The Protector Echo creates significant energy savings because it doesn't exhaust tempered room air from the lab and has no make-up air! It also eliminates air pollutant emissions.
The hood is constructed of a glacier white powder-coated steel exterior and a chemical-resistant, fiberglass-reinforced, composite panel liner. The vertical-rising sash is a single piece of tempered safety glass with cable pulley, sash weight and powder-coated aluminum sash handle and tracks with Clean-Sweep™ airflow openings. Sash stop is at 16" sash opening height. Sash opens to 28" high for loading. The sightline from the work surface and header panel is 37.5" (95.3 cm) high.
Certifications: All models conform or are certified to the following regulations and standards: UL 61010-1; CAN/CSA C22.2 No. 61010.1; SEFA 9-2010, DHIII; SEFA 8-2010, Cabinet Surface Finish Tests; ASHRAE 110-95; ANSI Z9.5-2011; AFNOR NF-X 15-211; ASTM E84-09C.
Accessories information: Radio Frequency Identification (RFID) Cards are required for use and maintenance providing additional safety and data/usage records by ensuring only authorized personnel access the hood.The Administration Card provides full access to view stored information on the fume hood. It is required to operate/view stored data on the fume hood, including user time usage.
Ordering information: The Protector Echo benchtop Filtered Fume Hood is available in 3 styles: no windows, side windows, and 360° windows, and 4 widths: 4', 5', 6', and 8'. Each hood requires a work surface, base cabinet or stand, and Neutrodine Filters (sold separately). The number of filters needed depends on the hood's width. Optional accessories include ADA Remote control, gGuard® Communication software, HEPA Filter, and Sash Opening Reduction Wing Kits.
NOTE: The Lifetime Payback Questionnaire (LPQ) form is REQUIRED for all Protector Echo Filtered Fume Hood orders. It must be completed in full, submitted, reviewed, and approved before an order can be processed. Lead times do not start until the LPQ is approved. Contact your sales representative or Labconco for more information or a copy of the form.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: VWR CATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST@vwr.com for more details. You can also visit https://vwr.com/equip_services.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty