Would you like to log in again?
Protector® Pass-Through Laboratory Hoods, Labconco®
Supplier: Labconco
113407002 113608002 113608000 113408020 113508021 113408021 113500000 113408002 113600020 113500020 113507002 113407021 113600021 113508020 113400002 113600000 113608021 113607000 113408000 113608020 113508002 113607020 113507021 113507000 113407020 113607021 113400020 113407000 113400000 113400021 113600002 113607002 113500021 113500002 113508000 113507020
10804-956EA 13625 USD InStock
10804-956 10799-248 10804-958 10804-970 10804-972 10805-098 10799-254 10799-232 10799-234 10799-256 10799-250 10799-230 10799-252 10804-966 10804-968 10799-236 10799-258 10799-214 10799-216 10799-238 10805-096 10804-960 10804-962 10804-940 10804-964 10804-942 10805-104 10799-260 10805-102 10805-100 10799-244 10799-246 10799-240 10799-262 10799-242 10805-106
Protector® Pass-Through Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Laboratory hoods feature sashes on two sides that, when mounted in an island or peninsular configuration, provide unobstructed viewing from either side during chemistry demonstrations or observing students using the hood
Ergonomic air foils allow air to sweep the work surface for maximum containment. Clean-Sweep* airflow openings pull inflow air from under the airfoil so that clean air continually flows over the airfoil, creating a constant barrier of protection from contaminants. Upper baffle efficiently directs airflow to the interior center. Bypass airflow design ensures relatively stable face velocities. The 25.6cm (103/32") I.D. exhaust connection is made of powder-coated steel.
Durable frame is constructed of powder-coated steel and aluminum. Chemical-resistant, fiberglass-reinforced composite panel liner surpasses all national codes for flame spread and has a bright white surface for excellent light reflectivity. Front and side panels may be easily removed for lamp replacement and access to sash weight and electrical supply connections. Clear, vertical-rising tempered safety glass sashes and streamlined corner posts provide maximum visibility. An optional 38.1Wx61Hcm (15x24") tempered, safety glass viewing window can be mounted on the left or right side for enhanced visibility. An incandescent light, switches, and a GFCI electrical duplex receptacle are factory pre-wired to the hood's junction box. Color: glacier white.
SpillStopper Work Surfaces (sold separately) are solid, black surfaces molded of a special formulation of corrosion-resistant epoxy resin. They are dished and contoured to conform to interior liner of Protector Pass-Through Hoods. Pre-drilled 2.5" (6.4cm) dia plumbing holes. May be ordered with pre-cut 6" x 3" (15.2 x 7.6 cm) oval cupsink cutout(s). Cupsink is sold separately.
These hoods may also be installed in the wall between two rooms. An audible alarm sounds when both sashes are opened at the same time.
Certifications: Hoods meet SEFA 1, ASHRAE 110-95, ANSI Z9.5, UL 3101-1/61010-1, and CAN/CSA C22.2 No. 1010.1 standards. ETL listed.
Ordering information: Hoods require use of a remote blower, ductwork, work surface, and base cabinet (all sold separately); contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: VWR CATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST@vwr.com for more details. You can also visit https://vwr.com/equip_services.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty