Would you like to log in again?
WORKSURFACE PT 6FT-LS RS CSK
Supplier: Labconco
SpillStopper^ SpillStopper, For 6ft Protector Pass-Through Laboratory Hood, Corrosion resistant epoxy resin, Pre-drilled, Dished, Contoured, Width: 6 ft, Thickness: 3.2cm, Dimension: 76.2D x 182.9cm W, Cupsink Cutout(s): Left side, right side
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WORKSURFACE PT 6FT-LS RS CSK 10805-988 9570622 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty