Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
RXPert™ Single Filtered Balance Systems, Labconco®
Supplier: Labconco
3971401 3971476 3971576 3971695 3971202 3971200 3971302 3971400 3971495 3971675 3971697 3971677 3971201 3971301 3971575 3971300 3971596 3971402 3971475 3971477 3971496 3971497 3971696 3971676 3971595 3971597 3971577
10830-818EA 20840 USD InStock
10830-818 10830-816 10216-860 10830-814 10830-812 10830-810 10216-856 10830-820 10830-786 10216-858 10216-852 10216-854 10830-808 10830-806 10830-826 10830-804 10216-850 10830-802 10830-824 10830-822 10830-800 10830-788 10216-846 10216-848 10830-792 10216-844 10830-790
RXPert™ Single Filtered Balance Systems, Labconco®
Fume Hoods and Enclosures Balances Enclosures
The patented RXPert Single Filtered Balance System is a Class I Enclosure that meets the USP<800> requirements for non-sterile compounding
The RXPert features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. This balance enclosure includes an all-in-one factory-installed canopy, and damper allows for thimble ducting to the outside. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement.
The HEPA filter is easily accessed to allow the entire surface to be scanned for leak detection. It includes a built-in blower with dynamically-balanced, vibration-isolated motorized impeller and speed control. The blower has a 40% reserve so that the speed may be adjusted as needed by a certified technician as the filter loads.
Optional filtration kits and thimble ducting kits are available to convert existing XPert Balance Filtered Systems, XPert Bulk Powder Filtered Systems, and RXPert Single Filtered Balance Systems. Double Filtered models requiring no ductwork are also available.
It provides user protection by keeping powders and particulates contained during hazardous drug manipulation. It features durable glass and steel construction, ergonomic air foil with aerodynamic Clean-Sweep airflow openings, and deep interior of 23.4" or 30.0" depending on the model.
Certifications: ETL listed and CE marked. All models conform to the following: SEFA 1-2010; Modified ASHRAE 110-1995; ANSI Z9.5-2011; UL 61010-1; CAN/CSA C22.2 No. 1010.1.
Ordering information: All models include a built-in analog airflow monitor. Widths of 2', 3' and 4' with standard height and 4’, 5’ and 6’ with tall height or tall height and extended depth are available. Some models include a stainless steel or epoxy work surface. A work surface is required if one is not included. All models require (not included): a remote blower, ductwork and supporting base.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty