Would you like to log in again?
MOLEC SIEVE REPLCMNT MEDIA 5LB
Supplier: Labconco
Molecular sieve replacement media 5lb^ Molecular sieve replacement media, for replacement spent media in drying train column. provides two refills. Glove box accessory.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MOLEC SIEVE REPLCMNT MEDIA 5LB 15202-532 7837300 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty