Would you like to log in again?
Inlet Filter, Labconco®
Supplier: Labconco
1472500 1472200
26671-506EA 963.13 USD InStock
26671-506 26671-516
Inlet Filter, Labconco®
Filters Liquid Process Filters
This disposable filter prevents oil backstreaming from the vacuum pump and protects the pump from damage.
Filter life is approximately 1000 hours at 10 µm vacuum.
Delivery information: Includes filter assembly and one filter cartridge.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty