Would you like to log in again?
Accessories for 4.5 L FreeZone® Freeze Dry Systems, Labconco
Supplier: Labconco
7537801 7353403 7762800 7756200 7537800
26671-774EA 478.93 USD InStock
26671-774 10148-050 10148-064 10148-052 26671-694
Accessories for 4.5 L FreeZone® Freeze Dry Systems, Labconco
Freeze Dryers
Attachment port lid has a 7.62 cm (3") diameter opening to permit the use of heated drying chamber or manifolds 26617-017, -021 or -023 atop the FreeZone® 4.5 L freeze dry systems (26671-511, -513).
Wire shelf racks attach easily without tools to the sides of the cabinet to provide additional support for glassware attached to the drying chamber. Charcoal epoxy-coated steel. Each shelf measures 17.8D x 49.2W cm (7 x 193/8").
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty