Would you like to log in again?
FreeZone® Freeze Dry Glassware, Labconco
Supplier: Labconco
FreeZone®
7558000 7572710 7574210 7557800 7554600 7568000 7568400 7571210 7570810 7573810 7568200 7550400 7573210 7554800 7550600 7573610 7557400 7557000 7555000 7552400 7556000 7557900 7556200 7552200 7557200 7554000 7570610 7557600 7550800 7552000 7554200 7574010 7554900 7554400 7556400 7571010 7550200 7575010 7568600 7573410 7550900 7573010 7550000 7575210 7551000
26674-526EA 238.94 USD InStock
26674-526 26675-090 26674-425 26674-524 26674-528 26674-440 26674-540 26674-522 26674-420 26674-145 26675-699 26674-140 30153-694 26675-691 26675-570 26675-572 26675-693 26675-079 26675-695 26675-574 26675-872 10148-042 26675-697 26675-576 26674-536 26674-415 26674-534 26674-538 26674-451 26674-350 26674-532 26674-214 26674-236 26674-156 26674-178 26674-530 26675-687 26675-104 26675-689 10148-034 26675-701 26674-250 26675-680 26675-068 26675-685
FreeZone® Freeze Dry Glassware, Labconco
Freeze Dryers
FreeZone® Freeze Dry Systems were designed to provide simplicity of use and convenience when freeze drying samples for the laboratory.
  • A variety of accessories, including glassware, are available for these units
  • Glassware offered includes flasks, ampules, serum bottles and vials
Labconco Ampules are fabricated of highest quality borosilicate glass for strength and durability
Serum Bottles are perfect for long term storage of freeze dried samples. Labconco serum bottles are specifically designed for lyophilization applications. Their uniform thin wall construction ensures even freezing and drying. Serum Bottles are ideal containers for use in the FreeZone Stoppering Tray Dryer and also connect to valve ports on drying chambers and manifolds.
Adapters connect Lyph-Lock® Flasks to the valve ports on a drying chamber or manifold of a freeze dryer. These adapters have a 90° bend.
Lyph-Lock® Flasks come in a variety of sizes to suit different sized samples. Flasks should be filled to no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is ensured. A complete Lyph-Lock Flask Flask includes a glass top and bottom and a rubber ring seal. Tops, bottoms and seals are available separately as replacement components. Adapters are required to attach flasks to freeze dry valve ports.
Ordering information: For Fast-Freeze Flasks in clear and amber-colored glass, contact your Avantor representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty