Would you like to log in again?
Aluminum Seals, Labconco®
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
7577110 7577010
26676-355PK 23.16 USD InStock
26676-355 26676-353
Aluminum Seals, Labconco®
Closures Chromatography Vial Closures
Aluminum Seal
Package of 100. Forms a tight seal around top of Split Stopper and lip of Serum Bottle when crimped. Specifically designed for lyophilization applications. For use with FreeZone Free Dry Systems. Seals are for serum bottles with 20 mm corkage or 13 mm corkage, as indicated.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty