Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goldfisch Fat and Oil Extractor, Labconco®

Supplier: Labconco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

3500100
27681-000EA 23220 USD InStock
27681-000
Goldfisch Fat and Oil Extractor, Labconco®
Extractors Automatic Extractor Systems
For determination of fat and oil content in samples not exceeding 5g

Reduces extraction time from 16 hours to approximately 4 hours. Frame is constructed of anodized aluminum. Extraction process involves locking any of 6 Alundum extraction thimbles to condenser and applying heat. Solvent condenses and washes down through sample into boiling solvent below. After extraction, solvent reclaiming tube replaces sample container. Solvent chamber is a beaker which is weighed before extraction begins. This beaker is also weighed after extraction and final solvent evaporation. The increase in beaker weight indicates the weight of fat or oil extracted.

A 3/8" IPS (International Pipe Standard) control valve on the upper left side is for connection to a water source. A 3/4" IPS water outlet on the upper right side is for connection to an open drain of at least 3/4" IPS. An on/off switch is lighted red when in the on position.

Listed in AOAC® methods for determining fat and oil. Exhaust flammable solvents inside an explosion-proof fume hood.

Uses solvents whose boiling range does not exceed 85°C.

Ordering information: Manufacturer's one-year warranty on parts and labor. Comes fully assembled. Includes 6' line cord with grounded plug. Supplied with six 100W heater elements, 6 stirrup supports, 6 stainless steel condensers, and 6 automatic pressure-release valves. Includes 12 thick-wall borosilicate glass 100 mL beakers with flanged, ground top. Also includes 6 Alundum extraction thimbles, 6 heat covers, 6 cork gaskets, 6 beaker rings, 6 upper condenser gaskets, 9 sample tubes, and 9 solvent reclaiming tubes.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty