Would you like to log in again?
HEPA FLTR CELLSF/CBN/HEPA ENCL
Supplier: Labconco
HEPA Color-Smart* Filter Cells^ For use with Paramount* Duet* and Duet 360* Filtered Enclosures. 99.99% efficient for particulates 0.3um or larger, such as bacteria and chemical powders.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HEPA FLTR CELLSF/CBN/HEPA ENCL 30140-274 6933200 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty