Would you like to log in again?
FLTR CELL ENCL F/AMMONI 1PR=PK
Supplier: Labconco
Ammonia Filter Cells^ Type CX, suitable for ammonia and low molecular weight amines. For Paramount* and Paramount Duet* Filtered Enclosures. Each cell contains a minimum of 11.8kg of impregnated carbon. Color-coded bright green. Pair includes three contaminant-detector tubes.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLTR CELL ENCL F/AMMONI 1PR=PK 30153-978 6910200 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty