Would you like to log in again?
FILTER ROUGHNG F/PARAMOUNT PK2
Supplier: Labconco
Roughing Prefilter^ Included on Paramount* and Paramount Duet* Filtered Enclosures when purchased. Removes dust and mist to extend the life of the filter cells. It is recommended that roughing prefilters be replaced when filter cells are replaced.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER ROUGHNG F/PARAMOUNT PK2 30153-993 6911000 Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty