Would you like to log in again?
DRYING CHAMBER HEATED 220V
Supplier: Labconco
Drying Chamber^ Heated. 230V, 50/60Hz. 23Diameterx27Hcm. For FreeZone* Freeze Dry Systems. Type 304 stainless steel. Three-shelf product heater with a +43[degree]C high limit thermostat and a 24V transformer. Includes clear acrylic cover, gaskets and electrical passthrough for transformer cords.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DRYING CHAMBER HEATED 220V 30181-302 7521001 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty