Would you like to log in again?
POLISH KIT WTRPRO PS/UF
Supplier: Labconco
UF Polishing Kit^ For WaterPro* PS/UF Polishing Stations (26301-857, -867, -866, -868). Includes one carbon cartridge, two deionization cartridges, two hollow fiber final filters for dispensing valve and gun, and an ultrafilter.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
POLISH KIT WTRPRO PS/UF 30181-419 9047401 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty