Would you like to log in again?
POLISHING KIT HPLC/UF
Supplier: Labconco
Polishing kit HPLC/UF^ Hybrid polishing kit for Waterpro PS HPLC/UF hybrid polishing stations 90007-04 & 90007-05. includes 1 carbon cartridge, 2 deionization cartridges and 1 organic adsorption cartridge. Water purif. supplies.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
POLISHING KIT HPLC/UF 30181-420 9066600 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty