Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
RapidVap® N₂ Evaporation System, Labconco®
Supplier: Labconco
RapidVap®
7913900 7910000 7925500 7914008 7910010 7913500 7913608 7926808 7913700 7925900 7926600 7925408 7926008 7913808 7926908
26676-046EA 171.57 USD InStock
26676-046 26676-057 26676-058 26676-044 26676-042 26676-054 26676-004 13500-080 13500-050 37001-384 13500-082 13500-052 30181-430 13500-054 13500-056
RapidVap® N₂ Evaporation System, Labconco®
Evaporators Centrifugal Evaporators
Perfect for fast, unattended concentration of environmental samples
System signals completion of a run in two ways. In one, a timer pauses the system and alerts user when the predetermined end point is achieved. In the second method, the system monitors the temperature differential between the block and heater. When these are nearly equal, meaning end point is near, the alarm sounds and light flashes. The unique Cool Zone™ block and tube design reduces the evaporation rate after end point is achieved.
The Cool Zone™ insulates the samples and allows the researcher adequate time to remove the samples before they reach dryness. System consists of a glacier white and charcoal epoxy-coated aluminum base with thermoplastic upper housing, a fluoropolymer resin-coated aluminum chamber, a nitrogen port, rear 5.1cm (2") diameter exhaust port, and a 1.3cm (1/2") thick glass lid with safety switch.
Microprocessor-controlled programming for vortex speed from 0 to 100%; heat from OFF to 100°C; time from 1 to 999 minutes; and number of active nitrogen positions, 2, 4, 6, or 8. Memory stores 1 to 9 programs. LCD screen displays program number, actual and set point vortex speed, actual and set point temperature, time remaining, and number of active nitrogen positions. Control panel has run/stop button, preheat/end alarm button and indicator light, set point selection buttons, and increase/decrease programming buttons.
Fluoropolymer resin-coated aluminum blocks are available for both 170 and 600mL sizes. Each graduated 600mL tube has a sample capacity of up to 450mL when vortexing. Each graduated 170mL tube has a sample capacity of up to 125mL when vortexing. Glassware caps help prevent cross contamination and moisture from condensing when using low boiling point solvents.
System combines mechanical vortex action up to 500rpm, heat, and nitrogen blow down technologies. Process eight samples up to 450mL in size at once. The microprocessor-controlled nitrogen blow down manifold directs nitrogen flow into the selected sample tubes.
Certifications: The 230V units are CE marked.
Ordering information: System includes 1 eight-place fluoropolymer resin-coated aluminum block for 600mL tubes, 8 polyethylene caps for 600mL tubes, and 1.8m (6') of 5.1cm (2") diameter exhaust ducting. The 115V models include a 2m (6'6") three-wire cord with 20A plug.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty