Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Elbows, Labconco

Supplier: Labconco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7027300 5605100 7027400 5605200 4719100 5602100 4708800 4708700 4719000
82006-930EA 1630 USD InStock
82006-930 82006-932 30182-144 30182-111 30182-136 30152-640 30182-124 30182-326 30182-290
Elbows, Labconco
Ducting Supplies
PVC elbows, both 45° and 90°, are compatible with thermoplastic duct

Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty