Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Canopy Hood, Labconco®
Supplier: Labconco
4799000 7099000
30192-104EA 3560 USD InStock
30192-104 30192-159
Canopy Hood, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Specifically designed to solve unique ventilation problems, this canopy hood vents nontoxic materials such as heat, steam, and odors from large or bulky equipment
Hood is shipped fully assembled with integral baffle and mounting hardware for peninsular or island locations*. Hood interior and exterior are epoxy-coated, cold-rolled steel. Requires remotely located blower for operation. Contact your Avantor Furniture Specialist for assistance with blower sizing.
*For ceiling mount, threaded rods (not included) may be required.
The epoxy-coated steel canopy has a built-in baffle to increase air velocities and enhance its overall capture efficiency. The canopy may be installed on a wall or suspended from the ceiling* for peninsula and corner locations. For most efficient performance, the canopy hood should be installed up to 30.5 cm (12") above the equipment to be ventilated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty